Assay Details
| Gene Symbol: Smc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133786.3 | NP_598547.1 | 17 - 18 | 2724 | 95 | |
| GenBank mRNA | AJ534940.2 | - | 16 - 17 | 2600 | 95 | |
| AK048650.1 | - | 12 - 13 | 1938 | 95 | ||
| BC061481.1 | - | 17 - 18 | 2699 | 6512832 | 95 | |
| BC062939.1 | - | 17 - 18 | 2724 | 6841276 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smc4
Genomic Map