Assay Details
| Gene Symbol: Fam168b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160236.1 | NP_001153708.1 | 3 - 4 | 383 | 68 | |
| NM_174997.2 | NP_778162.1 | 3 - 4 | 383 | 68 | ||
| GenBank mRNA | AK220453.1 | - | 3 - 4 | 383 | 68 | |
| BC040511.1 | - | 3 - 4 | 360 | 4977517 | 68 | |
| BC043098.1 | - | 3 - 4 | 307 | 6406013 | 68 | |
| BC059247.1 | - | 3 - 4 | 360 | 6409420 | 68 | |
| CA328263.1 | - | 2 - 3 | 131 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam168b
Genomic Map