Assay Details
| Gene Symbol: Fam168b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160235.1 | NP_001153707.1 | 6 - 7 | 736 | 63 | |
| NM_001160236.1 | NP_001153708.1 | 5 - 6 | 721 | 63 | ||
| NM_174997.2 | NP_778162.1 | 5 - 6 | 700 | 63 | ||
| GenBank mRNA | AK144298.1 | - | 5 - 6 | 600 | 63 | |
| AK220453.1 | - | 5 - 6 | 700 | 63 | ||
| BC040511.1 | - | 5 - 6 | 677 | 4977517 | 63 | |
| BC043098.1 | - | 5 - 6 | 624 | 6406013 | 63 | |
| BC059247.1 | - | 5 - 6 | 677 | 6409420 | 63 | |
| CA328263.1 | - | 4 - 5 | 469 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam168b
Genomic Map