Assay Details
| Gene Symbol: Nup153 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175749.2 | NP_786925.2 | 12 - 13 | 1904 | 59 | |
| GenBank mRNA | BC052173.1 | - | 12 - 13 | 1642 | 6516328 | 59 |
| CA326562.1 | - | 7 - 8 | 596 | 59 | ||
| CK638380.1 | - | 1 - 2 | 97 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nup153
Genomic Map