Assay Details
| Gene Symbol: Nup153 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175749.2 | NP_786925.2 | 14 - 15 | 2089 | 150 | |
| GenBank mRNA | BC046468.1 | - | 2 - 3 | 113 | 150 | |
| BC052173.1 | - | 14 - 15 | 1827 | 6516328 | 150 | |
| CK638380.1 | - | 3 - 4 | 282 | 150 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nup153
Genomic Map