Assay Details
| Gene Symbol: Dpep2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301204.1 | NP_001288133.1 | 9 - 10 | 1253 | 76 | |
| NM_001301205.1 | NP_001288134.1 | 8 - 9 | 938 | 76 | ||
| NM_176913.4 | NP_795887.2 | 10 - 11 | 1410 | 76 | ||
| GenBank mRNA | AF488552.1 | - | 10 - 11 | 1410 | 76 | |
| AK089244.1 | - | 9 - 10 | 1239 | 76 | ||
| AK155099.1 | - | 9 - 10 | 1205 | 76 | ||
| AK155308.1 | - | 9 - 10 | 1204 | 76 | ||
| AK155417.1 | - | 9 - 10 | 1208 | 76 | ||
| AK170094.1 | - | 9 - 10 | 1205 | 76 | ||
| AK170970.1 | - | 9 - 10 | 1205 | 76 | ||
| AK171044.1 | - | 9 - 10 | 1205 | 76 | ||
| AK171122.1 | - | 9 - 10 | 1205 | 76 | ||
| AK171155.1 | - | 8 - 9 | 918 | 76 | ||
| AK171196.1 | - | 9 - 10 | 1205 | 76 | ||
| AK171268.1 | - | 9 - 10 | 1205 | 76 | ||
| AK171503.1 | - | 9 - 10 | 1205 | 76 | ||
| AK171606.1 | - | 9 - 10 | 1221 | 76 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2067362 | AK171155.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2067362 | AK171268.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2067362 | AK171503.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2067362 | AK170970.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2067362 | AK171044.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2077368 | NM_176913.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2077368 | AF488552.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 117429 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 921783 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 640700 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 112480 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 462241 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 71270 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P101906 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P105494 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2077368 | NM_176913.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2077368 | AF488552.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26137225 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26137234 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26137224 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26137223 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26137215 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26137219 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26137237 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26137240 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26137222 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26137236 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26137220 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26137230 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26137216 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06628316 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P105494 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P101906 | - | - |
| Whole Mouse Genome G4122A | A_51_P209029 | - | - |
| Whole Mouse Genome G4122A | A_52_P502717 | AK171606.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P502717 | AK171155.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P502717 | AK171196.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P502717 | AK171268.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P502717 | AK171503.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108665 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P105494 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P101906 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06790746 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07182106 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06790758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137162 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137218 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137193 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137220 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137183 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137170 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07132711 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06880329 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07132715 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137212 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137243 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06804949 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137223 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07409755 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07409762 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07450980 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137196 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137230 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07409759 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137215 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137213 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07068271 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137222 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06796312 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137237 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137169 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137234 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06715255 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137200 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137167 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137238 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137163 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137191 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137172 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06790757 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07172411 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07409758 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137242 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137160 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137224 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07208891 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137206 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137166 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07450973 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137184 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06804950 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07336828 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07450968 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06894523 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137244 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07182106 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06790751 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137186 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137162 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137175 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137219 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07409761 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137159 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137208 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137225 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10581440 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581445 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581444 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581438 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581439 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581443 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581437 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581436 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581446 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581449 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581441 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581442 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10581448 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10581434 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3190364 | - | - |
| MouseWG-6 v2 Expression BeadChip | 106400278 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 104480452 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3190364 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106400278 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3130309 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003130309 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003190364 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1248177 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2692316 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2692316 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1248177 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2511213 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1234229 | - | - |
| mouse-6 v1.1 (Illumina) | scl000629.1_5-S | - | - |
| mouse-6 v1.1 (Illumina) | scl00319446.2_1-S | - | - |
| mouse-6 v1.1 (Illumina) | scl34381.6_178-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106400278 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 104480452 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3130309 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3190364 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl000629.1_5-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00319446.2_1-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104480452 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3190364 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106400278 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3130309 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2692315 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2692316 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2692316 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2511213 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1234229 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2692315 | - | - |

Dpep2
Genomic Map