Assay Details
| Gene Symbol: Dpep2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176913.4 | NP_795887.2 | 1 - 2 | 248 | 58 | |
| GenBank mRNA | AF488552.1 | - | 1 - 2 | 248 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dpep2
Genomic Map