Assay Details
| Gene Symbol: Gen1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177331.4 | NP_796305.3 | 7 - 8 | 1022 | 90 | |
| GenBank mRNA | AK030995.1 | - | 7 - 8 | 1032 | 90 | |
| AK031077.1 | - | 7 - 8 | 1001 | 90 | ||
| AK158255.1 | - | 6 - 7 | 845 | 90 | ||
| BC090653.1 | - | 7 - 8 | 1022 | 30659763 | 90 | |
| BC099936.1 | - | 6 - 7 | 831 | 90 | ||
| BC138157.1 | - | 7 - 8 | 909 | 8861176 | 90 | |
| BC138158.1 | - | 7 - 8 | 909 | 8861177 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gen1
Genomic Map