Assay Details
| Gene Symbol: Gen1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177331.4 | NP_796305.3 | 9 - 10 | 1212 | 95 | |
| GenBank mRNA | AK030995.1 | - | 9 - 10 | 1222 | 95 | |
| AK031077.1 | - | 9 - 10 | 1191 | 95 | ||
| AK158255.1 | - | 8 - 9 | 1035 | 95 | ||
| BC090653.1 | - | 9 - 10 | 1212 | 30659763 | 95 | |
| BC099936.1 | - | 8 - 9 | 1021 | 95 | ||
| BC138157.1 | - | 9 - 10 | 1099 | 8861176 | 95 | |
| BC138158.1 | - | 9 - 10 | 1099 | 8861177 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gen1
Genomic Map