Assay Details
| Gene Symbol: Hmbox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177338.5 | NP_796312.2 | 4 - 5 | 909 | 68 | |
| GenBank mRNA | AK052729.1 | - | 4 - 5 | 919 | 68 | |
| AK089782.1 | - | 4 - 5 | 910 | 68 | ||
| BC002212.1 | - | 2 - 3 | 313 | 68 | ||
| BC051457.1 | - | 4 - 5 | 912 | 6398463 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hmbox1
Genomic Map