Assay Details
| Gene Symbol: Hmbox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177338.5 | NP_796312.2 | 7 - 8 | 1263 | 64 | |
| GenBank mRNA | AK052729.1 | - | 7 - 8 | 1273 | 64 | |
| AK089782.1 | - | 7 - 8 | 1264 | 64 | ||
| BC002212.1 | - | 5 - 6 | 667 | 64 | ||
| BC051457.1 | - | 7 - 8 | 1266 | 6398463 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hmbox1
Genomic Map