Assay Details
| Gene Symbol: Lppr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177664.5 | NP_808332.3 | 3 - 4 | 1171 | 115 | |
| GenBank mRNA | AF541279.1 | - | 3 - 4 | 1172 | 115 | |
| AK046782.1 | - | 3 - 4 | 1172 | 115 | ||
| AK079635.1 | - | 3 - 4 | 505 | 115 | ||
| AK129149.1 | - | 3 - 4 | 767 | 115 | ||
| AY266266.1 | - | 3 - 4 | 1172 | 115 | ||
| AY266267.1 | - | 3 - 4 | 1172 | 115 | ||
| BC082323.1 | - | 3 - 4 | 701 | 115 | ||
| BC137701.1 | - | 3 - 4 | 766 | 8860720 | 115 | |
| BC137702.1 | - | 3 - 4 | 767 | 8860721 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lppr4
Genomic Map