Assay Details
| Gene Symbol: Nek5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177898.4 | NP_808566.2 | 6 - 7 | 547 | 64 | |
| GenBank mRNA | AK032672.1 | - | 6 - 7 | 549 | 64 | |
| AK054168.1 | - | 6 - 7 | 561 | 64 | ||
| AK165108.1 | - | 4 - 5 | 310 | 64 | ||
| BC053516.1 | - | 6 - 7 | 535 | 6311513 | 64 | |
More Information
Set Membership: |
|

Nek5
Genomic Map