Assay Details
| Gene Symbol: Dhx33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178367.4 | NP_848144.3 | 2 - 3 | 480 | 74 | |
| GenBank mRNA | AK035177.1 | - | 2 - 3 | 480 | 74 | |
| BC027842.1 | - | 2 - 3 | 477 | 74 | ||
| BC052172.1 | - | 2 - 3 | 474 | 6515901 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dhx33
Genomic Map