Assay Details
| Gene Symbol: Dhx33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178367.4 | NP_848144.3 | 5 - 6 | 1064 | 68 | |
| GenBank mRNA | AK035177.1 | - | 5 - 6 | 1064 | 68 | |
| BC027842.1 | - | 5 - 6 | 1061 | 68 | ||
| BC052172.1 | - | 5 - 6 | 1058 | 6515901 | 68 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Dhx33
Genomic Map