Assay Details
| Gene Symbol: Thnsl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178413.5 | NP_848500.3 | 2 - 3 | 157 | 61 | |
| GenBank mRNA | BC025604.1 | - | 2 - 3 | 120 | 5250319 | 61 |
| CB600010.1 | - | 2 - 3 | 127 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC025604.1, NM_178413.5 transcripts.
Set Membership: |
|

Thnsl2
Genomic Map