Assay Details
| Gene Symbol: Supt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178652.2 | NP_848767.1 | 2 - 3 | 354 | 74 | |
| GenBank mRNA | AK050321.1 | - | 2 - 3 | 201 | 74 | |
| AK075916.1 | - | 2 - 3 | 354 | 74 | ||
| AK087935.1 | - | 2 - 3 | 199 | 74 | ||
| AK089837.1 | - | 2 - 3 | 201 | 74 | ||
| BC051540.1 | - | 1 - 2 | 84 | 74 | ||
| BC096051.1 | - | 2 - 3 | 138 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Supt3
Genomic Map