Assay Details
| Gene Symbol: Supt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178652.2 | NP_848767.1 | 6 - 7 | 763 | 79 | |
| GenBank mRNA | AK075916.1 | - | 6 - 7 | 763 | 79 | |
| AK087935.1 | - | 6 - 7 | 608 | 79 | ||
| BC051540.1 | - | 5 - 6 | 493 | 79 | ||
| BC096051.1 | - | 6 - 7 | 547 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Supt3
Genomic Map