Assay Details
| Gene Symbol: Supt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178652.2 | NP_848767.1 | 8 - 9 | 954 | 81 | |
| GenBank mRNA | AK050321.1 | - | 8 - 9 | 801 | 81 | |
| AK075916.1 | - | 8 - 9 | 954 | 81 | ||
| AK087935.1 | - | 8 - 9 | 799 | 81 | ||
| BC051540.1 | - | 7 - 8 | 684 | 81 | ||
| BC096051.1 | - | 8 - 9 | 738 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Supt3
Genomic Map