Assay Details
| Gene Symbol: Zfp385b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113399.1 | NP_001106870.1 | 2 - 3 | 361 | 96 | |
| NM_001113400.1 | NP_001106871.1 | 2 - 3 | 383 | 96 | ||
| NM_178723.6 | NP_848838.2 | 4 - 5 | 893 | 96 | ||
| GenBank mRNA | AK046798.1 | - | 4 - 5 | 894 | 96 | |
| AK050293.1 | - | 2 - 3 | 384 | 96 | ||
| AK078551.1 | - | 2 - 3 | 336 | 96 | ||
| AK079322.1 | - | 4 - 5 | 776 | 96 | ||
| AK149352.1 | - | 4 - 5 | 874 | 96 | ||
| BC048543.1 | - | 2 - 3 | 361 | 96 | ||
| BC132352.1 | - | 2 - 3 | 472 | 40130629 | 96 | |
| BC138239.1 | - | 2 - 3 | 472 | 8861258 | 96 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Zfp385b
Genomic Map