Assay Details
| Gene Symbol: Zfp385b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113399.1 | NP_001106870.1 | 6 - 7 | 1056 | 114 | |
| NM_001113400.1 | NP_001106871.1 | 6 - 7 | 1078 | 114 | ||
| NM_178723.6 | NP_848838.2 | 8 - 9 | 1588 | 114 | ||
| GenBank mRNA | AK046798.1 | - | 8 - 9 | 1589 | 114 | |
| AK050293.1 | - | 6 - 7 | 1079 | 114 | ||
| AK078551.1 | - | 6 - 7 | 1033 | 114 | ||
| AK079322.1 | - | 8 - 9 | 1473 | 114 | ||
| AK149352.1 | - | 8 - 9 | 1569 | 114 | ||
| BC132352.1 | - | 6 - 7 | 1167 | 40130629 | 114 | |
| BC138239.1 | - | 6 - 7 | 1167 | 8861258 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp385b
Genomic Map