Assay Details
| Gene Symbol: Elmod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170691.1 | NP_001164162.1 | 6 - 7 | 667 | 68 | |
| NM_178736.5 | NP_848851.3 | 6 - 7 | 672 | 68 | ||
| GenBank mRNA | AK029044.1 | - | 6 - 7 | 643 | 68 | |
| AK036607.1 | - | 6 - 7 | 650 | 68 | ||
| AK036746.1 | - | 6 - 7 | 612 | 68 | ||
| AK041137.1 | - | 6 - 7 | 573 | 68 | ||
| AK048572.1 | - | 2 - 3 | 1878 | 68 | ||
| AK082730.1 | - | 5 - 6 | 439 | 68 | ||
| AK084950.1 | - | 6 - 7 | 672 | 68 | ||
| BC079654.1 | - | 6 - 7 | 633 | 6855187 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Elmod2
Genomic Map