Assay Details
Gene Symbol: AW551984 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199556.1 | NP_001186485.1 | 9 - 10 | 1252 | 66 | |
NM_178737.5 | NP_848852.1 | 9 - 10 | 1242 | 66 | ||
GenBank mRNA | AK028282.1 | - | 9 - 10 | 1216 | 66 | |
AK049098.1 | - | 9 - 10 | 1243 | 66 | ||
AK082321.1 | - | 9 - 10 | 1225 | 66 | ||
BC061211.1 | - | 8 - 9 | 938 | 30250574 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
