Assay Details
| Gene Symbol: Slc38a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178746.4 | NP_848861.1 | 12 - 13 | 1410 | 114 | |
| GenBank mRNA | AK043402.1 | - | 12 - 13 | 1398 | 114 | |
| AK044735.1 | - | 12 - 13 | 1400 | 114 | ||
| AK078926.1 | - | 6 - 7 | 650 | 114 | ||
| BC052361.1 | - | 12 - 13 | 1410 | 6705789 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc38a9
Genomic Map