Assay Details
| Gene Symbol: Slc38a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178746.4 | NP_848861.1 | 14 - 15 | 1665 | 89 | |
| GenBank mRNA | AK043402.1 | - | 14 - 15 | 1653 | 89 | |
| AK044735.1 | - | 14 - 15 | 1655 | 89 | ||
| AK078926.1 | - | 8 - 9 | 905 | 89 | ||
| BC052361.1 | - | 14 - 15 | 1665 | 6705789 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc38a9
Genomic Map