Assay Details
| Gene Symbol: Egflam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289496.1 | NP_001276425.1 | 14 - 15 | 2359 | 80 | |
| NM_001289498.1 | NP_001276427.1 | 14 - 15 | 2354 | 80 | ||
| NM_178748.6 | NP_848863.1 | 14 - 15 | 2359 | 80 | ||
| GenBank mRNA | AK031136.1 | - | 14 - 15 | 2268 | 80 | |
| AK033332.1 | - | 14 - 15 | 2276 | 80 | ||
| AK037223.1 | - | 14 - 15 | 2261 | 80 | ||
| AK041546.1 | - | 14 - 15 | 2240 | 80 | ||
| AK087659.1 | - | 14 - 15 | 2239 | 80 | ||
| AK158909.1 | - | 14 - 15 | 2355 | 80 | ||
| BC095994.1 | - | 14 - 15 | 2089 | 6467602 | 80 | |
| BC150710.1 | - | 14 - 15 | 2079 | 9087621 | 80 | |
| DQ223720.1 | - | 14 - 15 | 2063 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Egflam
Genomic Map