Assay Details
| Gene Symbol: Tmem72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178768.4 | NP_848883.2 | 3 - 4 | 386 | 113 | |
| GenBank mRNA | AK033856.1 | - | 3 - 4 | 522 | 113 | |
| AK049048.1 | - | 3 - 4 | 541 | 113 | ||
| AK049938.1 | - | 3 - 4 | 499 | 113 | ||
| AK085603.1 | - | 3 - 4 | 387 | 113 | ||
| AK141578.1 | - | 3 - 4 | 543 | 113 | ||
| BC093486.1 | - | 3 - 4 | 529 | 30697044 | 113 | |
| BC116441.1 | - | 3 - 4 | 370 | 40104616 | 113 | |
More Information
Set Membership: |
|

Tmem72
Genomic Map