Assay Details
| Gene Symbol: Bcorl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178782.4 | NP_848897.3 | 12 - 13 | 5089 | 59 | |
| GenBank mRNA | AK031119.1 | - | 8 - 9 | 1581 | 59 | |
| AK054259.1 | - | 8 - 9 | 1623 | 59 | ||
| AK081004.1 | - | 8 - 9 | 3657 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bcorl1
Genomic Map