Assay Details
| Gene Symbol: Trappc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161410.1 | NP_001154882.1 | 10 - 11 | 2134 | 69 | |
| NM_178811.4 | NP_848926.3 | 10 - 11 | 2360 | 69 | ||
| GenBank mRNA | AK049172.1 | - | 9 - 10 | 1394 | 69 | |
| AK132166.1 | - | 10 - 11 | 2146 | 69 | ||
| AK147985.1 | - | 10 - 11 | 2134 | 69 | ||
| BC002235.1 | - | 2 - 3 | 118 | 69 | ||
| BC024077.1 | - | 10 - 11 | 2303 | 5097580 | 69 | |
| BC030887.1 | - | 10 - 11 | 2128 | 4483611 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trappc12
Genomic Map