| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1200010885.mm.1 |
NM_001161411.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010885.mm.1 |
NM_001161411.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010885.mm.1 |
TC1200001569.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200010888.mm.1 |
NM_001161410.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010888.mm.1 |
NM_001161410.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010888.mm.1 |
NM_001161411.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010888.mm.1 |
NM_001161411.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010888.mm.1 |
NM_178811.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010888.mm.1 |
NM_178811.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010888.mm.1 |
TC1200001569.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200010889.mm.1 |
NM_001161410.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010889.mm.1 |
NM_001161410.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010889.mm.1 |
NM_001161411.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010889.mm.1 |
NM_001161411.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010889.mm.1 |
NM_178811.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010889.mm.1 |
NM_178811.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010889.mm.1 |
TC1200001569.mm.1 |
MTA TC |