Assay Details
| Gene Symbol: Trappc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161410.1 | NP_001154882.1 | 7 - 8 | 1858 | 88 | |
| NM_178811.4 | NP_848926.3 | 7 - 8 | 2084 | 88 | ||
| GenBank mRNA | AK049172.1 | - | 6 - 7 | 1118 | 88 | |
| AK132166.1 | - | 7 - 8 | 1870 | 88 | ||
| AK147985.1 | - | 7 - 8 | 1858 | 88 | ||
| BC024077.1 | - | 7 - 8 | 2027 | 5097580 | 88 | |
| BC030887.1 | - | 7 - 8 | 1852 | 4483611 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trappc12
Genomic Map