Assay Details
| Gene Symbol: Slc9b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178877.6 | NP_849208.4 | 7 - 8 | 1020 | 75 | |
| GenBank mRNA | AK029417.1 | - | 4 - 5 | 571 | 75 | |
| BC090977.1 | - | 7 - 8 | 973 | 30503721 | 75 | |
More Information
Set Membership: |
|

Slc9b2
Genomic Map