Assay Details
| Gene Symbol: Slc9b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178877.6 | NP_849208.4 | 11 - 12 | 1521 | 106 | |
| GenBank mRNA | AK029417.1 | - | 8 - 9 | 1072 | 106 | |
| AK039655.1 | - | 2 - 3 | 520 | 106 | ||
| BC090977.1 | - | 11 - 12 | 1474 | 30503721 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc9b2
Genomic Map