Assay Details
| Gene Symbol: Afap1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178928.4 | NP_849259.2 | 1 - 2 | 91 | 57 | |
| GenBank mRNA | AK035227.1 | - | 1 - 2 | 91 | 57 | |
| AK053714.1 | - | 1 - 2 | 90 | 57 | ||
| BC094505.1 | - | 1 - 2 | 40 | 57 | ||
| BC138923.1 | - | 1 - 2 | 52 | 8861945 | 57 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK035227.1, AK053714.1, BC138923.1, NM_178928.4 transcripts.
Set Membership: |
|

Afap1l1
Genomic Map