Assay Details
| Gene Symbol: Afap1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178928.4 | NP_849259.2 | 4 - 5 | 400 | 54 | |
| GenBank mRNA | AK035227.1 | - | 4 - 5 | 400 | 54 | |
| AK053714.1 | - | 4 - 5 | 399 | 54 | ||
| BC138923.1 | - | 4 - 5 | 361 | 8861945 | 54 | |
More Information
Updated Mapping Information
Set Membership: |
|

Afap1l1
Genomic Map