Assay Details
| Gene Symbol: Nostrin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181547.3 | NP_853525.3 | 7 - 8 | 595 | 121 | |
| GenBank mRNA | AJ564264.1 | - | 7 - 8 | 505 | 121 | |
| AY280963.1 | - | 7 - 8 | 548 | 121 | ||
| BC069942.1 | - | 7 - 8 | 572 | 30443942 | 121 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nostrin
Genomic Map