Assay Details
| Gene Symbol: Pik3r3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181585.5 | NP_853616.1 | 2 - 3 | 611 | 131 | |
| GenBank mRNA | AK132341.1 | - | 2 - 3 | 886 | 131 | |
| AK135693.1 | - | 2 - 3 | 920 | 131 | ||
| AK161822.1 | - | 2 - 3 | 879 | 131 | ||
| AK163413.1 | - | 2 - 3 | 885 | 131 | ||
| BC053102.1 | - | 2 - 3 | 611 | 6405534 | 131 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Citations (1)
Delineating biological pathways unique to embryonic stem cell-derived insulin-producing cell lines from their noninsulin-producing progenitor cell lines.
- Authors
- Chen TS, Tan SS, Yeo RW, Teh BJ, Luo R, Li G, Lim SK
- Journal
- Endocrinology - Page No 3600 (Sun Aug 01 00:00:00 EDT 2010)
- Volume (Year)
- 151 (2010)
- Product Usage
- -

Pik3r3
Genomic Map