Assay Details
| Gene Symbol: Pik3r3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181585.5 | NP_853616.1 | 5 - 6 | 1013 | 81 | |
| GenBank mRNA | AK031936.1 | - | 5 - 6 | 930 | 81 | |
| AK132341.1 | - | 5 - 6 | 1288 | 81 | ||
| AK135693.1 | - | 5 - 6 | 1322 | 81 | ||
| AK161822.1 | - | 5 - 6 | 1281 | 81 | ||
| AK163413.1 | - | 5 - 6 | 1287 | 81 | ||
| BC053102.1 | - | 5 - 6 | 1013 | 6405534 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pik3r3
Genomic Map