Assay Details
| Gene Symbol: Pik3r3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181585.5 | NP_853616.1 | 9 - 10 | 1585 | 83 | |
| GenBank mRNA | AK031936.1 | - | 9 - 10 | 1502 | 83 | |
| AK132341.1 | - | 9 - 10 | 1860 | 83 | ||
| AK135693.1 | - | 9 - 10 | 1894 | 83 | ||
| AK161822.1 | - | 9 - 10 | 1853 | 83 | ||
| BC053102.1 | - | 9 - 10 | 1585 | 6405534 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pik3r3
Genomic Map