Assay Details
| Gene Symbol: Rnls | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146342.2 | NP_001139814.2 | 3 - 4 | 670 | 76 | |
| NM_001167818.1 | NP_001161290.1 | 5 - 6 | 919 | 76 | ||
| GenBank mRNA | AK014866.1 | - | 5 - 6 | 788 | 76 | |
| AK018322.1 | - | 5 - 6 | 887 | 76 | ||
| AK170321.1 | - | 3 - 4 | 671 | 76 | ||
| BC052355.1 | - | 5 - 6 | 750 | 6770333 | 76 | |
| DQ788834.1 | - | 5 - 6 | 696 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rnls
Genomic Map