Assay Details
| Gene Symbol: Shq1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK012978.1 | - | 2 - 3 | 297 | 85 | |
| BC052657.1 | - | 2 - 3 | 289 | 30042589 | 85 | |
| BY776788.1 | - | 2 - 3 | 303 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Shq1
Genomic Map