Assay Details
| Gene Symbol: Dsg1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181680.1 | NP_859008.1 | 10 - 11 | 1517 | 158 | |
| GenBank mRNA | AK034104.1 | - | 1460 | 158 | ||
| AY192159.1 | - | 10 - 11 | 1517 | 158 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115906 | AY314983.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115906 | NM_181680.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115906 | AK034104.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115906 | AY192159.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 922390 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 667148 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 654201 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 609328 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 381792 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 763410 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 338878 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 656083 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 940360 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 209295 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 389388 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 411968 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 815147 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 440543 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 299182 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 390233 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 3486 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 536609 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 851279 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 121894 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 178917 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 117221 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 17210 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 69714 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 279501 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 860630 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 26917 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 97061 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 66858 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 57580 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 160535 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 90837 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140259 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P128985 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115906 | AY314983.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115906 | NM_181680.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115906 | AK034104.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115906 | AY192159.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P102432 | AK034104.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31538588 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31538592 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31538571 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31538581 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31538569 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31538566 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31538576 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31538574 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31538590 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P140259 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P128985 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03424610 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08930188 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03424604 | - | - |
| Whole Mouse Genome G4122A | A_52_P102432 | AK034104.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130164 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P128985 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P140259 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13819728 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13819726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31538806 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538481 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13823652 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538590 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538517 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538779 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538476 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538533 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538484 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13733649 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13637288 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13733647 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538489 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538393 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538472 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538408 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538573 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13806221 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13819726 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538610 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13665038 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538574 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13906176 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538390 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538395 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538607 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538780 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538492 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538465 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13950244 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538479 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538514 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538392 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538469 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538463 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538538 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538550 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13887273 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538413 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538587 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538531 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538488 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13665031 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13665033 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538409 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538376 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538772 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538417 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538603 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538460 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538762 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538578 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538458 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13640792 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538597 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13705079 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538769 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538511 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538524 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538394 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538774 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538486 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538411 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538509 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538380 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13760892 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538470 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538528 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538600 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538534 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13665030 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538468 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538467 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538494 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538459 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538592 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13655579 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538554 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538510 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538456 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538526 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538548 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13640795 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538569 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538482 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538407 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538581 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538384 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538523 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14083412 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538604 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538383 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538405 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13862392 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538558 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538389 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538556 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538406 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538375 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538474 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538536 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13760889 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31538588 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10454125 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454109 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454111 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454121 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454119 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454127 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454118 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454112 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454120 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454123 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454116 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454114 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454107 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454126 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454108 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454115 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454117 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454110 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454106 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10454113 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10454105 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1800001017.mm.1 | NM_181680.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001017.mm.1 | TC1800000152.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800001018.mm.1 | NM_181680.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001018.mm.1 | TC1800000152.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800001016.mm.1 | NM_181680.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001016.mm.1 | TC1800000152.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800001019.mm.1 | NM_181680.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001019.mm.1 | TC1800000152.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800001015.mm.1 | NM_181680.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001015.mm.1 | TC1800000152.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800001020.mm.1 | NM_181680.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001020.mm.1 | TC1800000152.mm.1 | MTA TC |
| Murine Genome U74v2 Set | 166420_at | - | - |
| Murine Genome U74v2 Set | 166420_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 545556 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6110075 | - | - |
| MouseWG-6 v2 Expression BeadChip | 106550239 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106550239 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6110075 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006110075 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2597795 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1253233 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2597795 | - | - |
| mouse-6 v1.1 (Illumina) | scl00211924.2_177-S | - | - |
| mouse-6 v1.1 (Illumina) | scl52215.16.1_306-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6110075 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106550239 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00211924.2_177-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106550239 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6110075 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2597795 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3001560 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2597795 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3001560 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1253233 | - | - |

Dsg1c
Genomic Map