Assay Details
| Gene Symbol: Mkl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181860.1 | NP_862908.1 | 3 - 4 | 322 | 68 | |
| GenBank mRNA | BC028459.1 | - | 3 - 4 | 322 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mkl2
Genomic Map