Assay Details
| Gene Symbol: Sdr16c5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181989.1 | NP_871789.1 | 3 - 4 | 692 | 100 | |
| GenBank mRNA | AK136508.1 | - | 2 - 3 | 663 | 100 | |
| AY277588.1 | - | 3 - 4 | 692 | 100 | ||
| BC104134.1 | - | 3 - 4 | 611 | 40045268 | 100 | |
| BC104135.1 | - | 3 - 4 | 611 | 40045269 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sdr16c5
Genomic Map