Assay Details
| Gene Symbol: Cox6a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007748.3 | NP_031774.1 | 2 | 160 | 66 | |
| GenBank mRNA | AK002402.1 | - | 2 | 162 | 66 | |
| AK019318.1 | - | 2 | 165 | 66 | ||
| AK166496.1 | - | 2 | 162 | 66 | ||
| BC003807.1 | - | 2 | 160 | 3487598 | 66 | |
| BC052816.1 | - | 2 | 166 | 6439987 | 66 | |
| L06465.1 | - | 2 | 159 | 66 | ||
| U08440.1 | - | 2 | 149 | 66 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cox6a1
Genomic Map