Assay Details
| Gene Symbol: Mdh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008617.2 | NP_032643.2 | 1 - 2 | 154 | 53 | |
| GenBank mRNA | AK002305.1 | - | 1 - 2 | 172 | 53 | |
| AK135162.1 | - | 1 - 2 | 167 | 53 | ||
| AK160553.1 | - | 1 - 2 | 137 | 53 | ||
| AK167809.1 | - | 1 - 2 | 137 | 53 | ||
| BC023482.1 | - | 1 - 2 | 154 | 5251192 | 53 | |
| M16229.1 | - | 1 - 2 | 113 | 53 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK002305.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mdh2
Genomic Map