Assay Details
| Gene Symbol: Klrb1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159902.1 | NP_001153374.1 | 5 | 1039 | 92 | |
| NM_010737.3 | NP_034867.3 | 6 | 1138 | 92 | ||
| GenBank mRNA | M77753.1 | - | 6 | 1037 | 91 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P42245 | M77676.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P42245 | M77753.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P42245 | NM_010737.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P42245 | NM_001159902.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P42245 | AF354262.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P42245 | M77676.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P42245 | M77753.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P42245 | NM_010737.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P42245 | NM_001159902.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P42245 | AF354262.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 438844 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 896812 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 421250 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 466750 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 951057 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 685726 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 135466 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P114318 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P172487 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P42245 | M77676.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P42245 | M77753.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P42245 | NM_010737.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P42245 | NM_001159902.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P42245 | AF354262.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P42245 | M77676.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P42245 | M77753.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P42245 | NM_010737.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P42245 | NM_001159902.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P42245 | AF354262.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24840130 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24840152 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24840140 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24840150 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24840146 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24840114 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24840120 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24840131 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24840142 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24840109 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24840132 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P172487 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06066326 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P114318 | - | - |
| Whole Mouse Genome G4122A | A_52_P42245 | M77676.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P42245 | M77753.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P42245 | NM_010737.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P42245 | NM_001159902.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P42245 | AF354262.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P42245 | M77676.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P42245 | M77753.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P42245 | NM_010737.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P42245 | NM_001159902.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P42245 | AF354262.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116309 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P172487 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P114318 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05673785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840140 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840128 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05673785 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05124545 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840142 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05480985 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840090 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05798525 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05149851 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840118 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840132 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840131 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840159 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840150 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840112 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05403656 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05798523 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840152 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05079513 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05539057 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840146 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840162 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840100 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05539052 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05552101 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05079508 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840140 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10548288 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548287 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548291 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548290 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548294 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548293 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548289 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548292 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450296_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10548286 | - | - |
| Mouse Genome 430 2.0 Array | 1450296_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1450296_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17057_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17057_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600025103.mm.1 | TC0600003210.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025103.mm.1 | NM_001159902.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025103.mm.1 | NM_001159902.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025103.mm.1 | NM_010737.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025103.mm.1 | NM_010737.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025106.mm.1 | TC0600003210.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025106.mm.1 | NM_001159902.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025106.mm.1 | NM_001159902.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025106.mm.1 | NM_010737.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025106.mm.1 | NM_010737.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025107.mm.1 | TC0600003210.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025107.mm.1 | NM_010737.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025107.mm.1 | NM_010737.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025105.mm.1 | TC0600003210.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025105.mm.1 | NM_001159902.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025105.mm.1 | NM_001159902.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025105.mm.1 | NM_010737.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025105.mm.1 | NM_010737.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025109.mm.1 | TC0600003210.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025109.mm.1 | NM_001159902.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025109.mm.1 | NM_001159902.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025109.mm.1 | NM_010737.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025109.mm.1 | NM_010737.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025101.mm.1 | TC0600003210.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025101.mm.1 | NM_001159902.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025101.mm.1 | NM_001159902.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025101.mm.1 | NM_010737.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025101.mm.1 | NM_010737.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025104.mm.1 | TC0600003210.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025104.mm.1 | NM_001159902.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025104.mm.1 | NM_001159902.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025104.mm.1 | NM_010737.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025104.mm.1 | NM_010737.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025108.mm.1 | TC0600003210.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025108.mm.1 | NM_001159902.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025108.mm.1 | NM_001159902.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025108.mm.1 | NM_010737.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025108.mm.1 | NM_010737.3 | Refseq |
| Mouse Expression Set 430 | 1450296_at | - | - |
| Mouse Expression Set 430 | 1450296_PM_at | - | - |
| Murine Genome U74v2 Set | 94772_PM_at | - | - |
| Murine Genome U74v2 Set | 94772_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450296_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450296_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450296_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450296_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 613430 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4120722 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4120722 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004120722 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2602983 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2602983 | - | - |
| mouse-6 v1.1 (Illumina) | scl28354.6_388-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4120722 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl28354.6_388-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4120722 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2602983 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2602983 | - | - |

Klrb1a
Genomic Map