Assay Details
| Gene Symbol: Rnase1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011271.2 | NP_035401.2 | 2 | 349 | 77 | |
| GenBank mRNA | AK075775.1 | - | 2 | 349 | 77 | |
| BC030036.1 | - | 2 | 329 | 1230895 | 77 | |
| M27814.1 | - | 2 | 316 | 77 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P249798 | NM_011271.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P249798 | AK019019.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P249798 | AK075775.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P249798 | NM_011271.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P249798 | AK019019.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P249798 | AK075775.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 278963 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P141593 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P249798 | NM_011271.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P249798 | AK019019.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P249798 | AK075775.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P249798 | NM_011271.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P249798 | AK019019.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P249798 | AK075775.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600339 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600319 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600307 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600346 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600315 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600326 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600341 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600334 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600342 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600327 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600332 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600330 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600312 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600344 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600347 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600349 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600323 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600313 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600324 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600337 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600317 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600329 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600357 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600311 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600321 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600335 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600360 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600305 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P141593 | - | - |
| Whole Mouse Genome G4122A | A_51_P472153 | NM_011271.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P472153 | AK019019.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P472153 | AK075775.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P472153 | BC030036.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P472153 | M27814.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P249798 | NM_011271.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P249798 | AK019019.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P249798 | AK075775.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P249798 | NM_011271.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P249798 | AK019019.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P249798 | AK075775.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112048 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129381 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P141593 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11685973 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11229805 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11229808 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11229803 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11229802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600329 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600319 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600356 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600300 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11229805 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600302 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11685972 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600304 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11127074 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600337 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600346 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600333 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11657100 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600357 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600327 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600326 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600347 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600329 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600351 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600372 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600362 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600364 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600330 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600292 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11229816 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600334 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600343 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600360 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600315 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11127079 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600359 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600305 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11229818 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600367 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600342 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11465937 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600349 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600323 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600312 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600311 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600293 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600299 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600313 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600294 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600324 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600298 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11685977 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11357819 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600317 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600296 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600295 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10419565 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10419564 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416523_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10419563 | - | - |
| Mouse Genome 430 2.0 Array | 1416523_at | - | - |
| Mouse Genome 430 2.0 Array | 1416523_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19752_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19752_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1400015038.mm.1 | NM_011271.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400015038.mm.1 | NM_011271.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400015038.mm.1 | TC1400002000.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400015035.mm.1 | NM_011271.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400015035.mm.1 | NM_011271.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400015035.mm.1 | TC1400002000.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400015036.mm.1 | NM_011271.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400015036.mm.1 | NM_011271.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400015036.mm.1 | TC1400002000.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400015037.mm.1 | NM_011271.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400015037.mm.1 | NM_011271.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400015037.mm.1 | TC1400002000.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416523_at | - | - |
| Mouse Expression Set 430 | 1416523_PM_at | - | - |
| Murine Genome U74v2 Set | 98041_at | - | - |
| Murine Genome U74v2 Set | 98041_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416523_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416523_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416523_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416523_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 907054 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6550364 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6550364 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006550364 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1220763 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1220763 | - | - |
| mouse-6 v1.1 (Illumina) | scl45606.2_68-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6550364 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl45606.2_68-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6550364 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1220763 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1220763 | - | - |

Rnase1
Genomic Map