Assay Details
| Gene Symbol: Prm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013637.4 | NP_038665.1 | 2 | 269 | 102 | |
| GenBank mRNA | AK006377.1 | - | 2 | 268 | 102 | |
| AK007068.1 | - | 2 | 269 | 102 | ||
| AK018980.1 | - | 2 | 272 | 102 | ||
| BC049599.1 | - | 268 | 6773822 | 102 | ||
| BC059733.1 | - | 2 | 268 | 6774637 | 102 | |
| K02926.1 | - | 2 | 254 | 102 | ||
| M27500.1 | - | 2 | 254 | 102 | ||
| X14003.1 | - | 2 | 268 | 102 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P533280 | BC059733.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P533280 | NM_013637.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P533280 | AK006377.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P533280 | AK007068.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P533280 | AK018980.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P533280 | BC059733.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P533280 | NM_013637.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P533280 | AK006377.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P533280 | AK007068.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P533280 | AK018980.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P156697 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P138847 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P163064 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P533280 | BC059733.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P533280 | NM_013637.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P533280 | AK006377.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P533280 | AK007068.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P533280 | AK018980.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P533280 | BC059733.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P533280 | NM_013637.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P533280 | AK006377.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P533280 | AK007068.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P533280 | AK018980.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P533287 | K02926.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P533287 | M27500.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P533287 | BC059733.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P533287 | X14003.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P533287 | NM_013637.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P533280 | BC059733.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P533280 | NM_013637.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P533280 | AK006377.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P533280 | AK007068.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P533280 | AK018980.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P533280 | BC059733.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P533280 | NM_013637.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P533280 | AK006377.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P533280 | AK007068.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P533280 | AK018980.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P162984 | K02926.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P162984 | M27500.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P162984 | BC059733.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P162984 | X14003.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P162984 | NM_013637.4 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30544538 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30544539 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30544536 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30544548 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30544544 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30544525 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30544541 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30544543 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30544531 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30544526 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30544547 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30544533 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30544529 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30544534 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P156697 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P138847 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P163064 | - | - |
| Whole Mouse Genome G4122A | A_51_P162984 | K02926.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P162984 | M27500.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P162984 | BC059733.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P162984 | X14003.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P162984 | NM_013637.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P533280 | BC059733.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P533280 | NM_013637.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P533280 | AK006377.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P533280 | AK007068.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P533280 | AK018980.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P533280 | BC059733.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P533280 | NM_013637.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P533280 | AK006377.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P533280 | AK007068.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P533280 | AK018980.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P533287 | K02926.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P533287 | M27500.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P533287 | BC059733.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P533287 | X14003.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P533287 | NM_013637.4 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107204 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P156697 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P163064 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P138847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544510 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30544553 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30544555 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30544533 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30544544 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30544525 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30544541 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30544543 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30544554 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30544540 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30544557 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30544538 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30544529 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30544546 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30544548 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30544526 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30544531 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30544536 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10437685 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437686 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439379_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1415955_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437054_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10437684 | - | - |
| Mouse Genome 430 2.0 Array | 1437054_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1439379_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1437054_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1439379_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1415955_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1415955_PM_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19118_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19118_at | - | - |
| Mouse Expression Set 430 | 1415955_x_at | - | - |
| Mouse Expression Set 430 | 1415955_PM_x_at | - | - |
| Mouse Expression Set 430 | 1437054_x_at | - | - |
| Mouse Expression Set 430 | 1439379_PM_x_at | - | - |
| Mouse Expression Set 430 | 1437054_PM_x_at | - | - |
| Mouse Expression Set 430 | 1439379_x_at | - | - |
| Murine Genome U74v2 Set | 99589_PM_f_at | - | - |
| Murine Genome U74v2 Set | 99589_f_at | - | - |
| Murine Genome U74v2 Set | 168579_PM_r_at | - | - |
| Murine Genome U74v2 Set | 168579_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1439379_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415955_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1437054_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1439379_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1437054_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415955_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1437054_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415955_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1437054_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1439379_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1439379_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415955_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 107040497 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 107040497 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2687652 | - | - |
| mouse-6 v1.1 (Illumina) | scl48764.4.1_9-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 107040497 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 107040497 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2806549 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2806549 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2687652 | - | - |

Prm1
Genomic Map