Assay Details
| Gene Symbol: Lyz2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK150998.1 | - | 4 | 1056 | 68 | |
| AK153244.1 | - | 4 | 1056 | 68 | ||
| AK159276.1 | - | 4 | 1103 | 68 | ||
| AK159640.1 | - | 4 | 1050 | 68 | ||
| BC002069.1 | - | 4 | 1045 | 3593360 | 68 | |
| BC019611.1 | - | 738 | 68 | |||
| BC054463.1 | - | 4 | 1045 | 6514977 | 68 | |
| Gene Symbol: Lyz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013590.4 | NP_038618.1 | 4 | 1054 | 68 | |
| GenBank mRNA | BC061129.1 | - | 4 | 1054 | 30296274 | 68 |
| CD742179.1 | - | 1 | 68 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2181738 | BC061129.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2181738 | CD742179.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2181738 | NM_013590.4 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 452153 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 954646 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 522955 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 174352 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2181738 | BC061129.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2181738 | CD742179.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2181738 | NM_013590.4 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27480425 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27480423 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27480433 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27480438 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27480415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27480407 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27480428 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27480435 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27480427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27480405 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27480413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27480439 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27480424 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27480418 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27480434 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27480429 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27480404 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27480431 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02228686 | - | - |
| Whole Mouse Genome G4122A | A_52_P582059 | BC061129.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P582059 | NM_013590.4 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08907178 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08422981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27480445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27480435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27480430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27480462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27480414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27480465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27480459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27480467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27480427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27480456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27480438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27480345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27480439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27480355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27480423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27480327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27480441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27480442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27480422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27480428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27480329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27480396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27480457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27480349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27480395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27480458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27480455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27480346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27480344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27480447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27480330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27480425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27480367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27480460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27480454 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27480418 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27480441 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27480398 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27480438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08657370 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27480448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27480434 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27480404 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27480423 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27480405 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08274576 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08215196 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27480414 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27480442 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27480427 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27480396 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08363781 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08747578 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27480449 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08747577 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27480435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27480387 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27480395 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27480416 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08363786 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27480407 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08363784 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27480447 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27480385 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27480415 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27480428 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27480439 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27480422 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27480390 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08477898 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27480445 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10372653 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10372654 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10372655 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436996_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439426_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450822_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10372652 | - | - |
| Mouse Genome 430 2.0 Array | 1439426_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1436996_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1439426_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1450822_at | - | - |
| Mouse Genome 430 2.0 Array | 1436996_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1450822_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000022135.mm.1 | TC1000002929.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000022144.mm.1 | NM_013590.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000022144.mm.1 | NM_013590.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000022144.mm.1 | TC1000002930.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000022147.mm.1 | NM_013590.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000022147.mm.1 | NM_013590.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000022147.mm.1 | TC1000002930.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000022146.mm.1 | NM_013590.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000022146.mm.1 | NM_013590.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000022146.mm.1 | TC1000002930.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000022145.mm.1 | NM_013590.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000022145.mm.1 | NM_013590.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000022145.mm.1 | TC1000002930.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1436996_PM_x_at | - | - |
| Mouse Expression Set 430 | 1439426_PM_x_at | - | - |
| Mouse Expression Set 430 | 1450822_at | - | - |
| Mouse Expression Set 430 | 1439426_x_at | - | - |
| Mouse Expression Set 430 | 1450822_PM_at | - | - |
| Mouse Expression Set 430 | 1436996_x_at | - | - |
| Murine Genome U74v2 Set | 162309_PM_at | - | - |
| Murine Genome U74v2 Set | 162309_at | - | - |
| Murine Genome U74v2 Set | 101753_s_at | - | - |
| Murine Genome U74v2 Set | 101753_PM_s_at | - | - |
| Murine Genome U74v2 Set | 139754_f_at | - | - |
| Murine Genome U74v2 Set | 139754_PM_f_at | - | - |
| Murine Genome U74v2 Set | 169724_PM_r_at | - | - |
| Murine Genome U74v2 Set | 169724_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1439426_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450822_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450822_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1439426_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1436996_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1436996_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1436996_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1439426_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1439426_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1436996_PM_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 454213 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6020195 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5700609 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6020195 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005700609 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006020195 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2736867 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1247933 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2736867 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1247933 | - | - |
| mouse-6 v1.1 (Illumina) | scl0017110.1_19-S | - | - |
| mouse-6 v1.1 (Illumina) | scl017110.1_293-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6020195 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5700609 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl017110.1_293-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0017110.1_19-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5700609 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6020195 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2878071 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2736867 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1247933 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2736867 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2878071 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1247933 | - | - |

Lyz2
Genomic Map